2,4-Difluor-3-hydroxybenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS36321G1 AS36321G5
ACTIAS36321G1EA 0 CHF
ACTIAS36321G1 ACTIAS36321G5
2,4-Difluor-3-hydroxybenzaldehyd
2,4-Difluoro-3-hydroxybenzaldehyde
Formel: C₇H₄F₂O₂
MDL Number: MFCD12405741
CAS-Nummer: 192927-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR