4-Fluor-3-nitrophenol

Lieferant: ACTIVATE SCIENTIFIC

AS31205G1 AS31205G5
ACTIAS31205G1EA 0 CHF
ACTIAS31205G1 ACTIAS31205G5
4-Fluor-3-nitrophenol
4-Fluor-3-nitrophenol
Formel: C₆H₄FNO₃
MW: 157.0993 g/mol
MDL Number: MFCD04038714
CAS-Nummer: 2105-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR