3-Chlor-5-hydroxybenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS36315G1 AS36315G5
ACTIAS36315G1EA 0 CHF
ACTIAS36315G1 ACTIAS36315G5
3-Chlor-5-hydroxybenzaldehyd
3-Chloro-5-hydroxybenzaldehyde
Formel: C₇H₅ClO₂
MW: 156,5664 g/mol
MDL Number: MFCD08234659
CAS-Nummer: 1829-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR