5-Nitro-m-kresol

Lieferant: ACTIVATE SCIENTIFIC

AS36687G1 AS36687G5
ACTIAS36687G1EA 0 CHF
ACTIAS36687G1 ACTIAS36687G5
5-Nitro-m-kresol
5-Nitro-m-kresol
Formel: C₇H₇NO₃
MDL Number: MFCD09038591
CAS-Nummer: 127818-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR