3-Amino-5-brom-2-methoxypyridin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
5-Bromo-2-methoxypyridin-3-amine
AS31641G1
AS31641G5
ACTIAS31641G1EA
0
CHF
ACTIAS31641G1
ACTIAS31641G5
3-Amino-5-brom-2-methoxypyridin
3-Amino-5-bromo-2-methoxypyridine
Formel:
C₆H₇BrN₂O MW: 203,04 g/mol |
MDL Number:
MFCD07368887 CAS-Nummer: 884495-39-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...