3-Amino-2-brom-5-methoxypyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Bromo-5-methoxypyridin-3-amine

AS36520G1 AS36520G5
ACTIAS36520G1EA 0 CHF
ACTIAS36520G1 ACTIAS36520G5
3-Amino-2-brom-5-methoxypyridin
3-Amino-2-bromo-5-methoxypyridine
Formel: C₆H₇BrN₂O
MW: 203,0366 g/mol
MDL Number: MFCD16659663
CAS-Nummer: 1043688-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR