2-Amino-5-brom-3-methoxypyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Bromo-3-methoxypyridin-2-amine

AS32728G1 AS32728G5
ACTIAS32728G1EA 0 CHF
ACTIAS32728G1 ACTIAS32728G5
2-Amino-5-brom-3-methoxypyridin
2-Amino-5-bromo-3-methoxypyridine
Formel: C₆H₇BrN₂O
MDL Number: MFCD09909849
CAS-Nummer: 42409-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR