(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP)
Lieferant: BACHEM BIOCHEMICA
Synonyms: 1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tris(dimethylamino)phosphonium hexafluorophosphate(V), BOP reagent, Benzotriazol-1-yloxytris(dimethylamino)-phosphonium hexafluorophosphate
4031234.0025 4031234.0100
BACE4031234.0025EA 0 CHF
BACE4031234.0025 BACE4031234.0100
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP)
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP)
Formel: C₁₂H₂₂F₆N₆OP₂
MW: 442.28 g/mol
MDL Number: MFCD00011948
CAS-Nummer: 56602-33-6
EINECS: 260-279-1