4-(Cyanmethoxy)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (4-(Cyanomethoxy)phenyl)boronic acid

AS15055G1 AS15055G5
ACTIAS15055G1EA 0 CHF
ACTIAS15055G1 ACTIAS15055G5
4-(Cyanmethoxy)phenylboronsäure
4-(Cyanmethoxy)phenylboronsäure
Formel: C₈H₈BNO₃
MW: 176.9700 g/mol
MDL Number: MFCD09908170
CAS-Nummer: 947533-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR