2-Cyan-4-methoxybenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS40225G1 AS40225G10 AS40225G5
ACTIAS40225G1EA 0 CHF
ACTIAS40225G1 ACTIAS40225G10 ACTIAS40225G5
2-Cyan-4-methoxybenzaldehyd
2-Cyano-4-methoxybenzaldehyde
Formel: C₉H₇NO₂
MDL Number: MFCD08275456
CAS-Nummer: 21962-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR