5-Brom-2-iodbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS31605G1 AS31605G5
ACTIAS31605G1EA 0 CHF
ACTIAS31605G1 ACTIAS31605G5
5-Brom-2-iodbenzaldehyd
5-Brom-2-iodbenzaldehyd
Formel: C₇H₄BrIO
MW: 310.9200 g/mol
MDL Number: MFCD07779026
CAS-Nummer: 689291-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR