2-Brom-4-iodbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS33759G1 AS33759G5
ACTIAS33759G1EA 0 CHF
ACTIAS33759G1 ACTIAS33759G5
2-Brom-4-iodbenzaldehyd
2-Bromo-4-iodobenzaldehyde
Formel: C₇H₄BrIO
MDL Number: MFCD07779025
CAS-Nummer: 261903-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR