3-(2-Methoxyphenyl)-1H-pyrazol-5-amin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Amino-3-(2-methoxyphenyl)-1H-pyrazole

AS20912G1 AS20912G5
ACTIAS20912G1EA 248 CHF
ACTIAS20912G1 ACTIAS20912G5
3-(2-Methoxyphenyl)-1H-pyrazol-5-amin
3-(2-Methoxyphenyl)-1H-pyrazol-5-amin
Formel: C₁₀H₁₁N₃O
MDL Number: MFCD02664277
CAS-Nummer: 149246-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR