(±)-2-Piperazincarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Piperazine-2-carboxylic acid

AS20218G100 AS20218G25
ACTIAS20218G100EA 103 CHF
ACTIAS20218G100 ACTIAS20218G25
(±)-2-Piperazincarbonsäure
(±)-2-Piperazincarbonsäure
Formel: C₅H₁₀N₂O₂
MW: 130,15 g/mol
MDL Number: MFCD00132878
CAS-Nummer: 2762-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR