2-Chlor-5-(trifluormethoxy)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS50118G1 AS50118G5
ACTIAS50118G1EA 0 CHF
ACTIAS50118G1 ACTIAS50118G5
2-Chlor-5-(trifluormethoxy)phenylboronsäure
2-Chlor-5-(trifluormethoxy)phenylboronsäure
Formel: C₇H₅BClF₃O₃
MDL Number: MFCD06656267
CAS-Nummer: 1022922-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR