(±)-1-Boc-3-hydroxypyrrolidin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
tert-Butyl 3-hydroxypyrrolidine-1-carboxylate, 3-Hydroxypyrrolidine, N-BOC protected
AS20044G100
AS20044G25
AS20044G5
ACTIAS20044G100EA
218
CHF
ACTIAS20044G100
ACTIAS20044G25
ACTIAS20044G5
(±)-1-Boc-3-hydroxypyrrolidin
(±)-1-Boc-3-hydroxypyrrolidin
Formel:
C₉H₁₇NO₃ MW: 187.24 g/mol |
MDL Number:
MFCD04038535 CAS-Nummer: 103057-44-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...