(±)-1-Boc-3-(methylamino)pyrrolidin

Lieferant: ACTIVATE SCIENTIFIC

AS31295G1 AS31295G5
ACTIAS31295G1EA 0 CHF
ACTIAS31295G1 ACTIAS31295G5
(±)-1-Boc-3-(methylamino)pyrrolidin
(±)-1-Boc-3-(methylamino)pyrrolidin
Formel: C₁₀H₂₀N₂O₂
MDL Number: MFCD06656606
CAS-Nummer: 454712-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR