2-(Trifluormethyl)nicotinsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Trifluoromethylnicotinic acid
AS35554G1
AS35554G5
ACTIAS35554G1EA
0
CHF
ACTIAS35554G1
ACTIAS35554G5
2-(Trifluormethyl)nicotinsäure
2-(Trifluormethyl)nicotinsäure
Formel:
C₇H₄F₃NO₂ MW: 191,11 g/mol |
MDL Number:
MFCD01862052 CAS-Nummer: 131747-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...