Methyl-6-hydroxynicotinat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 6-oxo-1,6-dihydro-3-pyridinecarboxylate, Methyl 6-hydroxynicotinate

AS35355G1 AS35355G5
ACTIAS35355G1EA 0 CHF
ACTIAS35355G1 ACTIAS35355G5
Methyl-6-hydroxynicotinat
Methyl-6-hydroxynicotinat
Formel: C₇H₇NO₃
MW: 153,14 g/mol
MDL Number: MFCD04113616
CAS-Nummer: 66171-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR