3-Nitropropionsäure, Calbiochem®

Lieferant: Merck Millipore Calbiochem

164603-50MGEA 195 CHF
164603-50MG
3-Nitropropionsäure, Calbiochem®
3-Nitropropionsäure
Formel: C₃H₅NO₄
MW: 119,08 g/mol
MDL Number: MFCD00007406
CAS-Nummer: 504-88-1
EINECS: 208-003-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR