Methyl-7-indolcarboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 1H-indole-7-carboxylate

AS50150G100 AS50150G5
ACTIAS50150G100EA 0 CHF
ACTIAS50150G100 ACTIAS50150G5
Methyl-7-indolcarboxylat
Methyl-7-indolcarboxylat
Formel: C₁₀H₉NO₂
MDL Number: MFCD00211064
CAS-Nummer: 93247-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR