Methyl-6-indolcarboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 1H-indole-6-carboxylate

AS50149G1 AS50149G5
ACTIAS50149G1EA 0 CHF
ACTIAS50149G1 ACTIAS50149G5
Methyl-6-indolcarboxylat
Methyl-6-indolcarboxylat
Formel: C₁₀H₉NO₂
MDL Number: MFCD00211063
CAS-Nummer: 50820-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR