1-Boc-5-cyan-2-indolboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (1-(tert-Butoxycarbonyl)-5-cyano-1H-indol-2-yl)boronic acid, 5-Cyano-1H-indole-2-boronic acid, N-BOC protected

AS36226G1 AS36226G5
ACTIAS36226G1EA 0 CHF
ACTIAS36226G1 ACTIAS36226G5
1-Boc-5-cyan-2-indolboronsäure
1-Boc-5-cyan-2-indolboronsäure
Formel: C₁₄H₁₅BN₂O₄
MW: 286.09 g/mol
MDL Number: MFCD04114578
CAS-Nummer: 475102-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR