4-Brom-2-chlorbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Chloro-4-bromobenzaldehyde

AS31182G1 AS31182G5
ACTIAS31182G1EA 0 CHF
ACTIAS31182G1 ACTIAS31182G5
4-Brom-2-chlorbenzaldehyd
4-Bromo-2-chlorobenzaldehyde
Formel: C₇H₄BrClO
MW: 219,46 g/mol
MDL Number: MFCD08741413
CAS-Nummer: 158435-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR