2-Amino-5-chlor-2'-fluorbenzophenon

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (2-Amino-5-chlorophenyl)(2-fluorophenyl)methanone, 2-Amino-5-chloro-2′-fluorobenzophenone

AS24815G100 AS24815G25
ACTIAS24815G100EA 0 CHF
ACTIAS24815G100 ACTIAS24815G25
2-Amino-5-chlor-2'-fluorbenzophenon
2-Amino-5-chlor-2'-fluorbenzophenon
Formel: C₁₃H₉ClFNO
MW: 249.67 g/mol
Siedepunkt: 364
Melting Pt: 94…95 °C
MDL Number: MFCD00038381
CAS-Nummer: 784-38-3
EINECS: 212-316-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR