4'-Chlorvalerophenon

Lieferant: ACTIVATE SCIENTIFIC

AS34440G10 AS34440G100
ACTIAS34440G10EA 0 CHF
ACTIAS34440G10 ACTIAS34440G100
4'-Chlorvalerophenon
4'-Chlorvalerophenon
Formel: C₁₁H₁₃ClO
Siedepunkt: 100 °C (0,5 mmHg)
Melting Pt: 31 °C
MDL Number: MFCD00018716
CAS-Nummer: 25017-08-7
EINECS: 246-566-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR