4'-Isopropylacetophenon

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: p-Isopropylacetophenone, 4-Isopropylacetophenone

AS23894G100 AS23894G25
ACTIAS23894G100EA 0 CHF
ACTIAS23894G100 ACTIAS23894G25
4'-Isopropylacetophenon
4'-Isopropylacetophenon
Formel: C₁₁H₁₄O
MW: 162,23 g/mol
Dichte: 0.97 g/cm³ (20 °C)
MDL Number: MFCD00048297
CAS-Nummer: 645-13-6
EINECS: 211-433-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR