2',4',6'-Trihydroxyacetophenon Monohydrat
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
Synonyms:
2-Acetylphloroglucinol monohydrate, THAP monohydrate, Phloroacetophenone monohydrate
AS20060G100
AS20060G25
ACTIAS20060G100EA
0
CHF
ACTIAS20060G100
ACTIAS20060G25
2',4',6'-Trihydroxyacetophenon Monohydrat
2',4',6'-Trihydroxyacetophenon Monohydrat
Formel:
C₈H₈O₄·1H₂O Siedepunkt: 394 °C (1013 hPa) Melting Pt: 216…218 °C |
MDL Number:
MFCD00149091 CAS-Nummer: 207-556-5 EINECS: 207-556-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...