4'-(Trifluormethyl)acetophenon

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 1-[4-(Trifluoromethyl)phenyl]ethan-1-one, 4-(Trifluoromethyl)acetophenone, p-(Trifluoromethyl)acetophenone

AS21475G100 AS21475G25
ACTIAS21475G100EA 171 CHF
ACTIAS21475G100 ACTIAS21475G25
4'-(Trifluormethyl)acetophenon
4'-(Trifluormethyl)acetophenon
Formel: C₉H₇F₃O
MW: 188,15 g/mol
Siedepunkt: 205 °C (1013 hPa)
Melting Pt: 26…29 °C
Dichte: 0,924 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00000401
CAS-Nummer: 709-63-7
EINECS: 211-913-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR