3',4',5'-Trifluoracetophenon

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-(3,4,5-Trifluorophenyl)ethan-1-one

AS33203G10 AS33203G5
ACTIAS33203G10EA 0 CHF
ACTIAS33203G10 ACTIAS33203G5
3',4',5'-Trifluoracetophenon
3',4',5'-Trifluoracetophenon
Formel: C₈H₅F₃O
MW: 174.12 g/mol
MDL Number: MFCD00083533
CAS-Nummer: 220141-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR