3,5-Dichlortoluol

Lieferant: ACTIVATE SCIENTIFIC

AS40273G1 AS40273G5
ACTIAS40273G1EA 0 CHF
ACTIAS40273G1 ACTIAS40273G5
3,5-Dichlortoluol
3,5-Dichlortoluol
Formel: C₇H₆Cl₂
MDL Number: MFCD00035819
CAS-Nummer: 25186-47-4
EINECS: 246-722-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR