6-Nitro-2,4-xylidin

Lieferant: ACTIVATE SCIENTIFIC

AS21731G25 AS21731G5
ACTIAS21731G25EA 218 CHF
ACTIAS21731G25 ACTIAS21731G5
6-Nitro-2,4-xylidin
6-Nitro-2,4-xylidin
Formel: C₈H₁₀N₂O₂
MDL Number: MFCD00007739
CAS-Nummer: 1635-84-3
EINECS: 216-662-0
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR