3'-(Trifluormethyl)acetophenon
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
Synonyms:
m-(Trifluoromethyl)acetophenone, 1-[3-(Trifluoromethyl)phenyl]ethan-1-one
AS20271G100
AS20271G25
ACTIAS20271G100EA
0
CHF
ACTIAS20271G100
ACTIAS20271G25
3'-(Trifluormethyl)acetophenon
3'-(Trifluormethyl)acetophenon
Formel:
C₉H₇F₃O MW: 188,15 g/mol Siedepunkt: 198 °C (1013 hPa) Dichte: 1,23 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00000391 CAS-Nummer: 349-76-8 EINECS: 206-490-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...