4'-tert-Butyl-2-chloracetophenon

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-(tert-Butyl)phenacyl chloride

AS35120G1 AS35120G10 AS35120G5
ACTIAS35120G1EA 0 CHF
ACTIAS35120G1 ACTIAS35120G10 ACTIAS35120G5
4'-tert-Butyl-2-chloracetophenon
4'-tert-Butyl-2-chloracetophenon
Formel: C₉H₉ClO
MDL Number: MFCD00171621
CAS-Nummer: 21886-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR