4-(4-Pyridyl)benzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS30258G1 AS30258G5
ACTIAS30258G1EA 0 CHF
ACTIAS30258G1 ACTIAS30258G5
4-(4-Pyridyl)benzaldehyd
4-(4-Pyridyl)benzaldehyd
Formel: C₁₂H₉NO
MDL Number: MFCD02684110
CAS-Nummer: 99163-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR