3-(4-Pyridyl)benzaldehyd
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-(Pyridin-4-yl)benzaldehyde, 3-Pyrid-4-ylbenzaldehyde
AS30266G1
AS30266G5
ACTIAS30266G1EA
0
CHF
ACTIAS30266G1
ACTIAS30266G5
3-(4-Pyridyl)benzaldehyd
3-(4-Pyridyl)benzaldehyd
Formel:
C₁₂H₉NO MW: 183,2060 g/mol |
MDL Number:
MFCD02684105 CAS-Nummer: 208190-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...