4-(3-Pyridyl)benzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-(Pyridin-3-yl)benzaldehyde

AS30257G1 AS30257G5
ACTIAS30257G1EA 0 CHF
ACTIAS30257G1 ACTIAS30257G5
4-(3-Pyridyl)benzaldehyd
4-(3-Pyridyl)benzaldehyd
Formel: C₁₂H₉NO
MW: 183,2060 g/mol
MDL Number: MFCD01631910
CAS-Nummer: 127406-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR