2-(3-Pyridyl)benzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS30260G1 AS30260G5
ACTIAS30260G1EA 0 CHF
ACTIAS30260G1 ACTIAS30260G5
2-(3-Pyridyl)benzaldehyd
2-(3-Pyridyl)benzaldehyd
Formel: C₁₂H₉NO
MDL Number: MFCD02684097
CAS-Nummer: 176690-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR