4-Methoxy-2-methylphenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-Methoxy-2-methylbenzeneboronic acid, (4-Methoxy-2-methylphenyl)boronic acid
AS51464G1
AS51464G5
ACTIAS51464G1EA
0
CHF
ACTIAS51464G1
ACTIAS51464G5
4-Methoxy-2-methylphenylboronsäure
4-Methoxy-2-methylphenylboronsäure
Formel:
C₈H₁₁BO₃ MW: 165.99 g/mol |
MDL Number:
MFCD02684315 CAS-Nummer: 208399-66-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...