2-Methoxy-5-methylphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Methoxy-5-methylbenzeneboronic acid

AS51463G1 AS51463G5
ACTIAS51463G1EA 0 CHF
ACTIAS51463G1 ACTIAS51463G5
2-Methoxy-5-methylphenylboronsäure
2-Methoxy-5-methylphenylboronsäure
Formel: C₈H₁₁BO₃
MW: 165,99 g/mol
MDL Number: MFCD02179465
CAS-Nummer: 127972-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR