3-Amino-5-brom-1H-pyrazol

Lieferant: ACTIVATE SCIENTIFIC

AS35676G1 AS35676G5
ACTIAS35676G1EA 0 CHF
ACTIAS35676G1 ACTIAS35676G5
3-Amino-5-brom-1H-pyrazol
3-Bromo-1H-pyrazol-5-amine
Formel: C₃H₄BrN₃
MW: 161,990 g/mol
MDL Number: MFCD18801156
CAS-Nummer: 1203705-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR