5-Chlor-2-fluornicotinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoro-5-chloropyridine-3-carboxylic acid

AS21313G1 AS21313G5
ACTIAS21313G1EA 142 CHF
ACTIAS21313G1 ACTIAS21313G5
5-Chlor-2-fluornicotinsäure
5-Chlor-2-fluornicotinsäure
Formel: C₆H₃ClFNO₂
MDL Number: MFCD04112518
CAS-Nummer: 884494-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR