2-Chlor-5-fluornicotinsäure

Lieferant: ACTIVATE SCIENTIFIC

AS31269G1 AS31269G5
ACTIAS31269G1EA 0 CHF
ACTIAS31269G1 ACTIAS31269G5
2-Chlor-5-fluornicotinsäure
2-Chlor-5-fluornicotinsäure
Formel: C₆H₃ClFNO₂
MW: 175,5449 g/mol
MDL Number: MFCD03092932
CAS-Nummer: 38186-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR