4,4-Dimethylcyclohexanol

Lieferant: ACTIVATE SCIENTIFIC

AS33646G1 AS33646G10 AS33646G5
ACTIAS33646G1EA 0 CHF
ACTIAS33646G1 ACTIAS33646G10 ACTIAS33646G5
4,4-Dimethylcyclohexanol
4,4-Dimethylcyclohexanol
Formel: C₈H₁₆O
MDL Number: MFCD00101954
CAS-Nummer: 932-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR