3,6-Dimethylsalicylaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS31001G1 AS31001G5
ACTIAS31001G1EA 0 CHF
ACTIAS31001G1 ACTIAS31001G5
3,6-Dimethylsalicylaldehyd
3,6-Dimethylsalicylaldehyd
Formel: C₉H₁₀O₂
MDL Number: MFCD04037328
CAS-Nummer: 1666-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR