4-Hydroxy-2,5-dimethylbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS31000G1 AS31000G5
ACTIAS31000G1EA 0 CHF
ACTIAS31000G1 ACTIAS31000G5
4-Hydroxy-2,5-dimethylbenzaldehyd
4-Hydroxy-2,5-dimethylbenzaldehyd
Formel: C₉H₁₀O₂
MW: 150,1745 g/mol
MDL Number: MFCD03411744
CAS-Nummer: 85231-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR