4-Fluor-3-nitrophenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Fluoro-3-nitrobenzeneboronic acid

AS36720G1 AS36720G5
ACTIAS36720G1EA 0 CHF
ACTIAS36720G1 ACTIAS36720G5
4-Fluor-3-nitrophenylboronsäure
4-Fluor-3-nitrophenylboronsäure
Formel: C₆H₅BFNO₄
MDL Number: MFCD08277236
CAS-Nummer: 352530-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR