4-Brom-2-(trifluormethoxy)benzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS32318G1 AS32318G5
ACTIAS32318G1EA 0 CHF
ACTIAS32318G1 ACTIAS32318G5
4-Brom-2-(trifluormethoxy)benzaldehyd
4-Bromo-2-(trifluoromethoxy)benzaldehyde
Formel: C₈H₄BrF₃O₂
MDL Number: MFCD12406886
CAS-Nummer: 220996-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR