2-Chlor-3-fluorphenol

Lieferant: ACTIVATE SCIENTIFIC

AS33585G1 AS33585G5
ACTIAS33585G1EA 0 CHF
ACTIAS33585G1 ACTIAS33585G5
2-Chlor-3-fluorphenol
2-Chlor-3-fluorphenol
Formel: C₆H₄ClFO
MW: 146.5500 g/mol
MDL Number: MFCD08166413
CAS-Nummer: 863870-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR