3-Amino-5-chlorbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS23518G1 AS23518G5
ACTIAS23518G1EA 94 CHF
ACTIAS23518G1 ACTIAS23518G5
3-Amino-5-chlorbenzonitril
3-Amino-5-chlorbenzonitril
Formel: C₇H₅ClN₂
MW: 152,58 g/mol
MDL Number: MFCD06797224
CAS-Nummer: 53312-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR